SY3H3D68052B Synthonix Corporation

Methyl 3-bromo-5-(hydroxymethyl)benzoate

  • MDL number No MDL found
  • Cas Number : 307353-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51979.html

Safety Information

MSDS https://synthonix.com/51979.html
Availability details for
Quantity
  • Availability
  • Add to Cart