SY3H3D68052E Synthonix Corporation

Methyl 4-bromo-2-hydroxy-6-methylbenzoate

  • Cas Number : 2089319-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51982.html

Safety Information

MSDS https://synthonix.com/51982.html
Availability details for
Quantity
  • Availability
  • Add to Cart