SY3H3D68052F Synthonix Corporation

Methyl 4-bromo-3-hydroxy-2-methylbenzoate

  • MDL number No MDL found
  • Cas Number : 1149388-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51983.html

Safety Information

MSDS https://synthonix.com/51983.html
Availability details for
Quantity
  • Availability
  • Add to Cart