SY3H3D680534 Synthonix Corporation

Methyl 2-fluoro-3-methoxybenzoate

  • MDL number No MDL found
  • Cas Number : 958991-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51993.html

Safety Information

MSDS https://synthonix.com/51993.html
Availability details for
Quantity
  • Availability
  • Add to Cart