SY3H3D68053A Synthonix Corporation

(6-Methoxybenzofuran-2-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 952737-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52005.html

Safety Information

MSDS https://synthonix.com/52005.html
Availability details for
Quantity
  • Availability
  • Add to Cart