SY3H3D680540 Synthonix Corporation

6-Methoxyindoline hydrochloride

  • MDL number MFCD09026837
  • Cas Number : 4770-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52102.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52102.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart