SY3H3D680547 Synthonix Corporation

2-(Methyl(phenyl)amino)acetic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 21911-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/52117.html

Safety Information

MSDS https://synthonix.com/52117.html
Availability details for
Quantity
  • Availability
  • Add to Cart