SY3H3D68054E Synthonix Corporation

(R)-N-Methyl-1-phenylethanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1096105-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/52164.html

Safety Information

MSDS https://synthonix.com/52164.html
Availability details for
Quantity
  • Availability
  • Add to Cart