SY3H3D680551 Synthonix Corporation

(S)-1-(3-Methoxyphenyl)ethanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1304771-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52175.html

Safety Information

MSDS https://synthonix.com/52175.html
Availability details for
Quantity
  • Availability
  • Add to Cart