SY3H3D680557 Synthonix Corporation

(R)-Methyl 2-amino-2-cyclohexylacetate hydrochloride

  • MDL number No MDL found
  • Cas Number : 14328-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/52203.html

Safety Information

MSDS https://synthonix.com/52203.html
Availability details for
Quantity
  • Availability
  • Add to Cart