SY3H3D68055B Synthonix Corporation

1-Methyl-1,2,3,4-tetrahydroquinoxaline dihydrochloride

  • MDL number No MDL found
  • Cas Number : 1259952-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52251.html

Safety Information

MSDS https://synthonix.com/52251.html
Availability details for
Quantity
  • Availability
  • Add to Cart