SY3H3D68055F Synthonix Corporation

6-Methylquinolin-7-amine

  • Cas Number : 129844-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52277.html

Safety Information

MSDS https://synthonix.com/52277.html
Availability details for
Quantity
  • Availability
  • Add to Cart