SY3H3D680561 Synthonix Corporation

3-Methyl-1-phenyl-1H-pyrazole

  • MDL number No MDL found
  • Cas Number : 1128-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52280.html

Safety Information

MSDS https://synthonix.com/52280.html
Availability details for
Quantity
  • Availability
  • Add to Cart