SY3H3D680563 Synthonix Corporation

4-Methyl-3-phenyl-1H-pyrazole

  • MDL number No MDL found
  • Cas Number : 13808-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52282.html

Safety Information

MSDS https://synthonix.com/52282.html
Availability details for
Quantity
  • Availability
  • Add to Cart