SY3H3D680567 Synthonix Corporation

7-Methoxyquinolin-3-amine

  • MDL number No MDL found
  • Cas Number : 87199-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/52288.html

Safety Information

MSDS https://synthonix.com/52288.html
Availability details for
Quantity
  • Availability
  • Add to Cart