SY3H3D68056B Synthonix Corporation

3-Methoxy-4-(oxazol-5-yl)aniline

  • MDL number No MDL found
  • Cas Number : 198821-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52305.html

Safety Information

MSDS https://synthonix.com/52305.html
Availability details for
Quantity
  • Availability
  • Add to Cart