SY3H3D68056C Synthonix Corporation

Methyl 2-(1H-indazol-3-yl)acetate

  • MDL number No MDL found
  • Cas Number : 131666-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/52306.html

Safety Information

MSDS https://synthonix.com/52306.html
Availability details for
Quantity
  • Availability
  • Add to Cart