SY3H3D680570 Synthonix Corporation

Methyl 1-methyl-1H-benzo[d]imidazole-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 131020-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52323.html

Safety Information

MSDS https://synthonix.com/52323.html
Availability details for
Quantity
  • Availability
  • Add to Cart