SY3H3D680572 Synthonix Corporation

Methyl 4-methylpyrazolo[1,5-a]pyridine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 127717-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52326.html

Safety Information

MSDS https://synthonix.com/52326.html
Availability details for
Quantity
  • Availability
  • Add to Cart