SY3H3D680574 Synthonix Corporation

Methyl 4-methoxy-1H-indazole-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 865887-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/52333.html

Safety Information

MSDS https://synthonix.com/52333.html
Availability details for
Quantity
  • Availability
  • Add to Cart