SY3H3D680578 Synthonix Corporation

Methyl 2-amino-5-methyl-4-(trifluoromethyl)benzoate

  • Cas Number : 872624-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52379.html

Safety Information

MSDS https://synthonix.com/52379.html
Availability details for
Quantity
  • Availability
  • Add to Cart