SY3H3D680579 Synthonix Corporation

2-Methoxyquinoline-3-boronic acid

  • Cas Number : 886853-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52382.html

Safety Information

MSDS https://synthonix.com/52382.html
Availability details for
Quantity
  • Availability
  • Add to Cart