SY3H3D68057A Synthonix Corporation

(2-(Methoxycarbonyl)-1H-indol-5-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 284660-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52385.html

Safety Information

MSDS https://synthonix.com/52385.html
Availability details for
Quantity
  • Availability
  • Add to Cart