SY3H3D68057B Synthonix Corporation

Methyl 4-acetamido-5-chloro-2-hydroxybenzoate

  • Cas Number : 24190-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52387.html

Safety Information

MSDS https://synthonix.com/52387.html
Availability details for
Quantity
  • Availability
  • Add to Cart