SY3H3D68057D Synthonix Corporation

6-Methoxy-1H-indene

  • MDL number No MDL found

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/52406.html

Safety Information

MSDS https://synthonix.com/52406.html
Availability details for
Quantity
  • Availability
  • Add to Cart