SY3H3D680586 Synthonix Corporation

Methyl bicyclo[4.2.0]octa-1,3,5-triene-7-carboxylate

  • MDL number No MDL found
  • Cas Number : 35095-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/52443.html

Safety Information

MSDS https://synthonix.com/52443.html
Availability details for
Quantity
  • Availability
  • Add to Cart