SY3H3D680587 Synthonix Corporation

3-(4-(Methylthio)phenyl)acrylic acid

  • Cas Number : 102016-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52447.html

Safety Information

MSDS https://synthonix.com/52447.html
Availability details for
Quantity
  • Availability
  • Add to Cart