SY3H3D68058E Synthonix Corporation

Methyl 2-(3-formylphenyl)acetate

  • MDL number No MDL found
  • Cas Number : 142327-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52459.html

Safety Information

MSDS https://synthonix.com/52459.html
Availability details for
Quantity
  • Availability
  • Add to Cart