SY3H3D68058F Synthonix Corporation

Methyl 2-(4-formylphenyl)acetate

  • MDL number No MDL found
  • Cas Number : 96524-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52460.html

Safety Information

MSDS https://synthonix.com/52460.html
Availability details for
Quantity
  • Availability
  • Add to Cart