SY3H3D680590 Synthonix Corporation

1-(4-Methoxyphenyl)propane-1,2-dione

  • MDL number No MDL found
  • Cas Number : 10557-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52461.html

Safety Information

MSDS https://synthonix.com/52461.html
Availability details for
Quantity
  • Availability
  • Add to Cart