SY3H3D680598 Synthonix Corporation

Methyl 1,4-Benzodioxane-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 3663-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52482.html

Safety Information

MSDS https://synthonix.com/52482.html
Availability details for
Quantity
  • Availability
  • Add to Cart