SY3H3D6805A0 Synthonix Corporation

(1-Methyl-5-phenyl-1H-pyrazol-4-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 1858252-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52515.html

Safety Information

MSDS https://synthonix.com/52515.html
Availability details for
Quantity
  • Availability
  • Add to Cart