SY3H3D6805A3 Synthonix Corporation

4-(2-Methylimidazol-1-yl)phenylamine

  • MDL number No MDL found
  • Cas Number : 74852-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52527.html

Safety Information

MSDS https://synthonix.com/52527.html
Availability details for
Quantity
  • Availability
  • Add to Cart