SY3H3D6805AA Synthonix Corporation

8-Methyl-3,4-dihydroisoquinolin-1(2H)-one

  • MDL number No MDL found
  • Cas Number : 1082041-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52554.html

Safety Information

MSDS https://synthonix.com/52554.html
Availability details for
Quantity
  • Availability
  • Add to Cart