SY3H3D6805AC Synthonix Corporation

Methyl 3-(2-aminophenyl)acrylate

  • Cas Number : 1664-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52562.html

Safety Information

MSDS https://synthonix.com/52562.html
Availability details for
Quantity
  • Availability
  • Add to Cart