SY3H3D6805AD Synthonix Corporation

1-Methylindoline-4-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 168899-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/52565.html

Safety Information

MSDS https://synthonix.com/52565.html
Availability details for
Quantity
  • Availability
  • Add to Cart