SY3H3D6805AE Synthonix Corporation

Methyl (E)-3-(3-aminophenyl)acrylate

  • Cas Number : 163487-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52568.html

Safety Information

MSDS https://synthonix.com/52568.html
Availability details for
Quantity
  • Availability
  • Add to Cart