SY3H3D6805B0 Synthonix Corporation

7-Methoxy-3,4-dihydroquinolin-2(1H)-one

  • MDL number No MDL found
  • Cas Number : 22246-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52571.html

Safety Information

MSDS https://synthonix.com/52571.html
Availability details for
Quantity
  • Availability
  • Add to Cart