SY3H3D6805B4 Synthonix Corporation

4-Methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine-7-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 141103-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52582.html

Safety Information

MSDS https://synthonix.com/52582.html
Availability details for
Quantity
  • Availability
  • Add to Cart