SY3H3D6805B7 Synthonix Corporation

Methyl 3,4-dihydro-2H-benzo[b][1,4]oxazine-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 121591-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52595.html

Safety Information

MSDS https://synthonix.com/52595.html
Availability details for
Quantity
  • Availability
  • Add to Cart