SY3H3D6805BC Synthonix Corporation

Methyl 2-(2-bromoethyl)benzoate

  • MDL number No MDL found
  • Cas Number : 25109-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52619.html

Safety Information

MSDS https://synthonix.com/52619.html
Availability details for
Quantity
  • Availability
  • Add to Cart