SY3H3D6805C1 Synthonix Corporation

Methyl 3-(4-bromophenyl)propanoate

  • MDL number No MDL found
  • Cas Number : 75567-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52626.html

Safety Information

MSDS https://synthonix.com/52626.html
Availability details for
Quantity
  • Availability
  • Add to Cart