SY3H3D6805C2 Synthonix Corporation

Methyl 3-bromo-2,4-dimethylbenzoate

  • Cas Number : 151859-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52627.html

Safety Information

MSDS https://synthonix.com/52627.html
Availability details for
Quantity
  • Availability
  • Add to Cart