SY3H3D6805C5 Synthonix Corporation

Methyl 5-bromo-2,4-dimethylbenzoate

  • Cas Number : 152849-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52630.html

Safety Information

MSDS https://synthonix.com/52630.html
Availability details for
Quantity
  • Availability
  • Add to Cart