SY3H3D6805C7 Synthonix Corporation

Methyl 4-ethyl-3-iodobenzoate

  • Cas Number : 51885-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52651.html

Safety Information

MSDS https://synthonix.com/52651.html
Availability details for
Quantity
  • Availability
  • Add to Cart