SY3H3D6805CA Synthonix Corporation

Methyl 2-(bromomethyl)-5-methoxybenzoate

  • MDL number No MDL found
  • Cas Number : 788081-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52656.html

Safety Information

MSDS https://synthonix.com/52656.html
Availability details for
Quantity
  • Availability
  • Add to Cart