SY3H3D6805CE Synthonix Corporation

Methyl 2-iodo-4,5-dimethoxybenzoate

  • MDL number No MDL found
  • Cas Number : 173043-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52663.html

Safety Information

MSDS https://synthonix.com/52663.html
Availability details for
Quantity
  • Availability
  • Add to Cart