SY3H3D6805D4 Synthonix Corporation

5-Morpholinopicolinic acid

  • MDL number No MDL found
  • Cas Number : 1072103-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52705.html

Safety Information

MSDS https://synthonix.com/52705.html
Availability details for
Quantity
  • Availability
  • Add to Cart