SY3H3D6805D5 Synthonix Corporation

Methyl 3-amino-3-(4-nitrophenyl)propanoate

  • MDL number No MDL found
  • Cas Number : 273920-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52709.html

Safety Information

MSDS https://synthonix.com/52709.html
Availability details for
Quantity
  • Availability
  • Add to Cart