SY3H3D6805D6 Synthonix Corporation

N-(4-Methoxy-2-methyl-6-nitrophenyl)acetamide

  • MDL number No MDL found
  • Cas Number : 500562-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52710.html

Safety Information

MSDS https://synthonix.com/52710.html
Availability details for
Quantity
  • Availability
  • Add to Cart